missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzoxazole 98.0+%, TCI America™

Catalog No. M24541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24541G 1 g
M24545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24541G Supplier TCI America Supplier No. M24541G

Chemical Identifiers

CAS 13673-62-6
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00160027
InChI Key CBXAWZGFEDZKFR-UHFFFAOYSA-N
PubChem CID 4178773
IUPAC Name 2-methylsulfanyl-1,3-benzoxazole
SMILES CSC1=NC2=CC=CC=C2O1

Specifications

Color Yellow
Boiling Point 122°C
Quantity 1 g
Formula Weight 165.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)benzoxazole