missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrothiophene 98.0+%, TCI America™

Numéro de catalogue. B025710ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B025710ML 10 mL
B025725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B025710ML Fournisseur TCI America Code fournisseur B025710ML
Il en reste null

Identifiants chimiques

CAS 1795-09-1
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00015539
InChI Key AJPGNQYBSTXCJE-UHFFFAOYNA-N
Synonym 2-methyltetrahydrothiophene, thiophene, tetrahydro-2-methyl, tetrahydro-2-methylthiophene, thiophene, tetrahydromethyl, 2-methylthiophane, 2-methylthi-olane, pubchem12832, 2-methylthiacyclopentane, acmc-209efp, 2-methyl tetrahydrothiophene
PubChem CID 15711
IUPAC Name 2-methylthiolane
SMILES CC1CCCS1

Spécifications

Color Yellow
Boiling Point 132°C
Quantity 10 mL
UN Number 1993
Formula Weight 102.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyltetrahydrothiophene