missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M18415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18415G Supplier TCI America Supplier No. M18415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57124-87-5
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD01632596
InChI Key DBPHPBLAKVZXOY-UHFFFAOYSA-N
Synonym 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, tetrahydro-2-methyl-3-furanthiol, 3-furanthiol, tetrahydro-2-methyl, 2-methyl-3-mercaptotetrahydrofuran, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, 3-mercapto-2-methyltetrahydrofuran, pubchem9972, ksc201q4p, 2-methyl-3-tetrahydrofuran thiol
PubChem CID 62128
IUPAC Name 2-methyloxolane-3-thiol
SMILES CC1C(CCO1)S

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 3336
Boiling Point 168°C
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture)