missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)phenol, 98%

Catalog No. AC455750050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455750050 5 g
AC455750010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455750050 Supplier Thermo Scientific Chemicals Supplier No. 455750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27489-33-4 | C7H8O3S | 172.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27489-33-4
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.2
InChI Key ZWTZHHHSRDUVRT-UHFFFAOYSA-N
PubChem CID 598998
IUPAC Name 2-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=CC=C1O

Specifications

Melting Point 88.0°C to 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula HOC6H4SO2CH3
Quantity 5 g
Synonym 2-methylsulfonyl phenol, 2-methanesulfonylphenol, 2-methylsulfonyl benzenol, phenol, 2-methylsulfonyl, phenol,2-methylsulfonyl, phenol, o-methylsulfonyl, methylsulfonylphenol, methylsulfonylbenzenol, acmc-1cbus, 2-methanesulfonyl-phenol
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material 2-(Methylsulfonyl)phenol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only