missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)benzeneboronic acid, 98%

Catalog No. AAH5244506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5244506 5 g
AAH5244503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5244506 Supplier Thermo Scientific Chemicals Supplier No. H5244506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 330804-03-0 | C7H9BO4S | 200.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330804-03-0
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.015
MDL Number MFCD02179472
InChI Key MSQFFCRGQPVQRS-UHFFFAOYSA-N
Synonym 2-methylsulfonyl phenylboronic acid, 2-methylsulfonyl phenyl boronic acid, 2-methylsulfonylphenyl boronic acid, 2-methanesulfonyl phenylboronic acid, 2-methylsulphonyl phenylboronic acid, 2-methanesulfonylphenylboronic acid, 2-methylsulphonyl benzeneboronic acid, 2-methylsulfonyl benzeneboronic acid, b-2-methylsulfonyl phenyl-boronic acid
PubChem CID 2773539
IUPAC Name (2-methylsulfonylphenyl)boronic acid
SMILES B(C1=CC=CC=C1S(=O)(=O)C)(O)O

Specifications

Melting Point 78°C to 84°C
Quantity 5 g
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 2-(Methylsulfonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only