missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazine 98.0+%, TCI America™

Catalog No. M073625ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M073625ML 25 mL
M0736250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M073625ML Supplier TCI America Supplier No. M073625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-08-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006142
InChI Key CAWHJQAVHZEVTJ-UHFFFAOYSA-N
Synonym methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
PubChem CID 7976
IUPAC Name 2-methylpyrazine
SMILES CC1=NC=CN=C1

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 135°C
Quantity 25 mL
UN Number 1993
Formula Weight 94.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylpyrazine
RTECSNumber UQ3675000
TSCA Yes