missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazine Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019552
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019552 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019552 Supplier Merck Emd Millipore Supplier No. PHR1223500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 2-Methylpyrazine is one of the major flavor volatiles reported in rice cakes, roasted peanuts and soybean products.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 109-08-0
Density 1.03 g/mL (at 25°C)
Physical Form Neat
Linear Formula C5H6N2
MDL Number MFCD00006142
Melting Point -29°C
Boiling Point 135°C (761 mmHg)
Quantity 500 mg
Formula Weight 94.11
Grade Certified Reference Material