missing translation for 'onlineSavingsMsg'
Learn More

2-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M13135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13135G Supplier TCI America Supplier No. M13135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16419-60-6
Molecular Formula C7H9BO2
MDL Number MFCD00093526
InChI Key NSJVYHOPHZMZPN-UHFFFAOYSA-N
PubChem CID 2733267
IUPAC Name (2-methylphenyl)boronic acid

Specifications

Quantity 5 g
Synonym 2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl
SMILES CC1=CC=CC=C1B(O)O
Molecular Weight (g/mol) 135.96
Formula Weight 135.96
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber ED7777777
TSCA No