missing translation for 'onlineSavingsMsg'
Learn More

(2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. M29075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29075G 5 g
M29071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29075G Supplier TCI America Supplier No. M29075G

Chemical Identifiers

CAS 210823-54-4
Molecular Formula C17H18F3IO3S
Molecular Weight (g/mol) 486.288
MDL Number MFCD20264884
InChI Key QFMYCPNBAGXLDL-UHFFFAOYSA-M
Synonym Mesityl(o-tolyl)iodonium Triflate
PubChem CID 71311052
IUPAC Name (2-methylphenyl)-(2,4,6-trimethylphenyl)iodanium;trifluoromethanesulfonate
SMILES CC1=CC=CC=C1[I+]C2=C(C=C(C=C2C)C)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 486.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate