missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019744
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019744 5 mL
111018165 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019744 Supplier Merck Emd Millipore Supplier No. 683105ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Methylpentane is a type of branched alkane classified under the volatile organic compounds (VOCs) family of chemicals.

Featured Industries: Petroleum

Specifications

CAS 107-83-5
Ignition Point 583°F
Vapor Pressure 6.77 psi (37.7°C)
Vapor Density 3 (vs. air)
Density 0.653 g/mL (at 25°C)
Flash Point -7°C - closed cup
Physical Form Neat
Linear Formula CH3 CH2 CH2 CH(CH3)2
MDL Number MFCD00009406
Melting Point -154°C
Boiling Point 62°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 86.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less