missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Méthylindole 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 60°C |
| Color | Blanc-jaune |
| Boiling Point | 272°C |
| Quantity | 25 g |
| Formula Weight | 131.18 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 2-Methylindole |
Sécurité et manipulation
| EINECSNumber | (5)-0131 |
| RTECSNumber | NM0345000 |
| TSCA | Yes |