missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Méthylhexane 99,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 90°C |
| Quantity | 5 mL |
| UN Number | 3295 |
| Formula Weight | 100.21 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-Methylhexane |
Sécurité et manipulation
| EINECSNumber | (2)-0007 |
| RTECSNumber | MO3871500 |
| TSCA | Yes |