missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptane 98.0+%, TCI America™

Catalog No. M05195G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05195G Supplier TCI America Supplier No. M05195G

Chemical Identifiers

CAS 592-27-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009518
InChI Key JVSWJIKNEAIKJW-UHFFFAOYSA-N
Synonym heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
PubChem CID 11594
IUPAC Name 2-methylheptane
SMILES CCCCCC(C)C

Specifications

Color Colorless
Boiling Point 117°C
Quantity 5 g
UN Number 1262
Formula Weight 114.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylheptane
EINECSNumber (2)-0008
TSCA No