missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Méthylène-1,3-propanediol 96,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 95°C |
| Quantity | 5 g |
| Synonym | 2-methylene-1,3-propanediol, 2-methylenepropane-1,3-diol, 1,3-propanediol, 2-methylene, 1,3-dihydroxy-2-methylenepropane, 1,3-dihydroxy-2-methylidenepropane, dimethylol ethylene, 2-methylene1,3-propanediol, 2-methylenepropan-1,3-diol, ksc573o4t, 1,3-propanediol,2-methylene |
| Formula Weight | 88.11 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-Methylene-1,3-propanediol |
Sécurité et manipulation
| TSCA | Yes |