missing translation for 'onlineSavingsMsg'
Learn More

2-(Methyldithio)isobutyraldehyde, 95%

Catalog No. AC265150050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC265150050 5 g Glass bottle
AC265150010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC265150050 Supplier Thermo Scientific Chemicals Supplier No. 265150050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67952-60-7 | C5H10OS2 | 150.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67952-60-7
Molecular Formula C5H10OS2
Molecular Weight (g/mol) 150.25
InChI Key VLBWEJJOETYCSE-UHFFFAOYSA-N
Synonym 2-methyldithio isobutyraldehyde, 2-methyl-2-methyldithio propanal, unii-3d5g4az6l5, propanal, 2-methyl-2-methyldithio, 2-methyldithio-isobutyraldehyde, alpha-methyldithio isobutyraldehyde, 2-methyl-2-methyldisulfanyl propanal, 2-methyl-2-methyldithio propionaldehyde, 2-methyldisulfanyl isobutyraldehyde, cocoa propanal
PubChem CID 106217
IUPAC Name 2-methyl-2-(methyldisulfanyl)propanal
SMILES CC(C)(C=O)SSC

Specifications

Color Yellow
Density 1.1000g/mL
Boiling Point 46.0°C (1.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5230
Quantity 5 g
Specific Gravity 1.1
Solubility Information Solubility in water: practically insoluble
Formula Weight 150.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(Methyldithio)isobutyraldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 267-914-1

RUO – Research Use Only