missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutylamine (contains 3-Methylbutylamine) 85.0+%, TCI America™

Catalog No. M07675ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07675ML 5 mL
M076725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07675ML Supplier TCI America Supplier No. M07675ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-15-1
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008147
InChI Key VJROPLWGFCORRM-UHFFFAOYSA-N
Synonym 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
PubChem CID 7283
IUPAC Name 2-methylbutan-1-amine
SMILES CCC(C)CN

Specifications

Color Yellow
Quantity 5 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutylamine (contains 3-Methylbutylamine)
EINECSNumber (2)-0133
TSCA Yes