missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylamine, 98%

Catalog No. AAL0524314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0524314 25 g
AAL0524306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0524314 Supplier Thermo Scientific Chemicals Supplier No. L0524314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-93-0 | C8H11N | 121.183 g/mol

2-Methylbenzylamine, is used in the preparation of boc-2-alkylanilines: N- (tert-butoxycarbonyl)-1-methylbenzylamine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzylamine, is used in the preparation of boc-2-alkylanilines: N- (tert-butoxycarbonyl)-1-methylbenzylamine

Solubility
Very soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008112
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN

Specifications

Density 0.979
Boiling Point 198°C to 200°C
Flash Point 83°C (181°F)
Refractive Index 1.544
Quantity 25 g
UN Number UN2735
Beilstein 1099406
Sensitivity Air Sensitive
Solubility Information Very soluble in water.
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 2-Methylbenzylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 201-952-1
RTECSNumber DP5800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only