missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzeneboronic acid, 98%

Catalog No. AAB2315414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2315414 25 g
AAB2315403 1 g
AAB2315406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2315414 Supplier Thermo Scientific Chemicals Supplier No. B2315414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16419-60-6 | C7H9BO2 | 135.96 g/mol

2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 16419-60-6
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00093526
InChI Key NSJVYHOPHZMZPN-UHFFFAOYSA-N
Synonym 2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl
PubChem CID 2733267
IUPAC Name (2-methylphenyl)boronic acid
SMILES CC1=CC=CC=C1B(O)O

Specifications

Melting Point ∼162°C to 164°C
Quantity 25 g
Beilstein 2935796
Solubility Information Slightly soluble in water.
Formula Weight 135.96
Percent Purity 98%
Chemical Name or Material 2-Methylbenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber ED7777777
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only