missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)ethanol 99.0+%, TCI America™

Catalog No. M0144500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0144500ML 500 mL
M014425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0144500ML Supplier TCI America Supplier No. M0144500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00002839
InChI Key OPKOKAMJFNKNAS-UHFFFAOYSA-N
Synonym 2-methylamino ethanol, n-methylethanolamine, methylethanolamine, ethanol, 2-methylamino, methylethylolamine, n-methylaminoethanol, n-methylmonoethanolamine, n-monomethylethanolamine, 2-hydroxyethyl methylamine, monomethylaminoethanol
PubChem CID 8016
ChEBI CHEBI:21763
IUPAC Name 2-(methylamino)ethan-1-ol
SMILES CNCCO

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 156°C
Quantity 500 mL
UN Number 2735
Formula Weight 75.11
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Methylamino)ethanol
EINECSNumber (2)-0295
RTECSNumber KL6650000
TSCA Yes