missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroquinoline 98.0+%, TCI America™

Catalog No. M14991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14991G Supplier TCI America Supplier No. M14991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 613-30-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00051736
InChI Key DXDPHHQJZWWAEH-UHFFFAOYSA-N
Synonym 6-nitroquinaldine, quinoline, 2-methyl-6-nitro, maybridge1_001441, acmc-1bb88, 1-2-aminoethyl pyrrole-2,5-dione,hydrochloride
PubChem CID 69172
IUPAC Name 2-methyl-6-nitroquinoline
SMILES CC1=CC=C2C=C(C=CC2=N1)[N+]([O-])=O

Specifications

Melting Point 168°C
Quantity 1 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitroquinoline