missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. CC14904CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC14904CB 250mg
CC14904DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC14904CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC14904CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 321309-42-6
Formule moléculaire C12H10O2
Poids moléculaire (g/mol) 186.21
Numéro MDL MFCD02681964
Clé InChI QRBXYIYZWZGINC-UHFFFAOYSA-N
Synonyme 2-methyl-5-phenyl-3-furaldehyde, 2-methyl-5-phenyl-furan-3-carbaldehyde, 2-methyl-5-phenyl-3-furancarboxaldehyde, 3-furancarboxaldehyde,2-methyl-5-phenyl, 3-furancarboxaldehyde, 2-methyl-5-phenyl
PubChem CID 2795313
Nom de l’IUPAC 2-methyl-5-phenylfuran-3-carbaldehyde
SOURIRES CC1=C(C=C(O1)C2=CC=CC=C2)C=O

Spécifications

Nom ou substance chimique 2-Methyl-5-phenyl-3-furaldehyde
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 186.21
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.