missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-ethylthiazoline 88.0+%, TCI America™

Catalog No. M068925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M068925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M068925ML Supplier TCI America Supplier No. M068925ML
Only null left

Chemical Identifiers

CAS 4293-61-2
Molecular Formula C6H11NS
Molecular Weight (g/mol) 129.221
MDL Number MFCD00015468
InChI Key LENVRVUQOXYSDH-UHFFFAOYSA-N
Synonym 4-Ethyl-2-methylthiazoline
PubChem CID 107254
IUPAC Name 4-ethyl-2-methyl-4,5-dihydro-1,3-thiazole
SMILES CCC1CSC(=N1)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 129.22
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-4-ethylthiazoline