missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(4-morpholinyl)benzenamine, 97%

Catalog No. AC432460050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432460050 5 g Glass bottle
AC432460010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432460050 Supplier Thermo Scientific Chemicals Supplier No. 432460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 581-00-0 | C11H16N2O | 192.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 581-00-0
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD10686817
InChI Key ZGJUJDQANIYVAL-UHFFFAOYSA-N
Synonym 2-methyl-4-morpholin-4-yl aniline, 2-methyl-4-morpholinoaniline, 2-methyl-4-4-morpholinyl aniline, 4-p-amino-m-tolyl morpholine, 4-4-morpholinyl-2-methylaniline, 2-methyl4-4-morpholinyl benzenamine, n-4-amino-3-methylphenyl morpholine, 2-methyl-4-4-morpholinyl benzenamine, 2-methyl-4-morpholin-4-yl-phenylamine, benzenamine, 2-methyl-4-4-morpholinyl
PubChem CID 21955000
IUPAC Name 2-methyl-4-morpholin-4-ylaniline
SMILES CC1=C(C=CC(=C1)N2CCOCC2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 92°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 192.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-(4-morpholinyl)benzenamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only