missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-trifluoromethylaniline 99.0+%, TCI America™

Catalog No. M13285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13285G 5 g
M13281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13285G Supplier TCI America Supplier No. M13285G

Chemical Identifiers

CAS 54396-44-0
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00153216
InChI Key TWLDBACVSHADLI-UHFFFAOYSA-N
Synonym 2-methyl-3-trifluoromethyl aniline, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, 3-amino-2-methyl benzotrifluoride, 2-methyl-3-trifluoromethyl-phenylamine, 2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, benzenamine, 2-methyl-3-trifluoromethyl
PubChem CID 2735932
IUPAC Name 2-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=CC=C1N)C(F)(F)F

Specifications

Melting Point 40°C
Quantity 5 g
Formula Weight 175.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-trifluoromethylaniline
EINECSNumber (3)-4077
TSCA No