missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-phenyl-1-propene, 99%

Catalog No. AAB2037014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2037014 25 g
AAB2037006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2037014 Supplier Thermo Scientific Chemicals Supplier No. B2037014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3290-53-7 | C10H12 | 132.206 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slighlty soluble in aqeous solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 3290-53-7
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00039815
InChI Key MXTNFIYGTWARIN-UHFFFAOYSA-N
Synonym 2-methyl-3-phenyl-1-propene, methallylbenzene, benzene, 2-methyl-2-propenyl, benzene, 2-methylallyl, 2-methylprop-2-en-1-yl benzene, 2-methyl-2-propenyl benzene, methallyl benzene, 2-methylallylbenzene, 2-benzyl-1-propene, 2-methylallyl benzene
PubChem CID 18687
IUPAC Name 2-methylprop-2-enylbenzene
SMILES CC(=C)CC1=CC=CC=C1

Specifications

Quantity 25 g
Solubility Information Slighlty soluble in aqeous solvents.
Formula Weight 132.21
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-phenyl-1-propene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only