missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-pentanone 98.0+%, TCI America™

Catalog No. M12495ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12495ML 5 mL
M124925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12495ML Supplier TCI America Supplier No. M12495ML

Chemical Identifiers

CAS 565-69-5
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009314
InChI Key HYTRYEXINDDXJK-UHFFFAOYSA-N
Synonym 2-methyl-3-pentanone, ethyl isopropyl ketone, 3-pentanone, 2-methyl, 4-methyl-3-pentanone, isopropyl ethyl ketone, 2-methyl-3-pentanal, iso-c3h7coc2h5, acmc-209lte, ksc272m0l
PubChem CID 11265
IUPAC Name 2-methylpentan-3-one
SMILES CCC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 mL
UN Number 1224
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-pentanone
EINECSNumber (2)-2475
TSCA No