missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-pentanol, 97+%

Numéro de catalogue. p-7044740
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0801509 10 g
AAL0801518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0801509 Fournisseur Thermo Scientific Chemicals Code fournisseur L0801509
Il en reste null

CAS: 565-67-3 | C6H14O | 102.177 g/mol

2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water

Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Identifiants chimiques

CAS 565-67-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004567
InChI Key ISTJMQSHILQAEC-UHFFFAOYSA-N
Synonym 2-methyl-3-pentanol, 3-pentanol, 2-methyl, propanol, 1-isopropyl, acmc-20anqw, 4-methyl-3-pentanol, 2-methyl-pentan-3-ol, 4-01-00-01716 beilstein handbook reference, istjmqshilqaec-uhfffaoysa
PubChem CID 11264
IUPAC Name 2-methylpentan-3-ol
SMILES CCC(C(C)C)O

Spécifications

Density 0.83
Boiling Point 127°C to 129°C
Flash Point 46°C (114°F)
Refractive Index 1.417
Quantity 10 g
UN Number UN2282
Beilstein 1718952
Solubility Information Amalgamate with ethanol,ethyl ether,benzene. Insoluble in water
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-pentanol
Afficher plus de résultats Afficher moins de résultats
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber 209-286-3
RTECSNumber SA7400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only