missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrophenylacetic Acid 96.0+%, TCI America™

Catalog No. M18175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18175G Supplier TCI America Supplier No. M18175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23876-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD07782132
InChI Key LWIOFILTAJJDLA-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrophenylacetic acid, 2-methyl-3-nitrophenyl acetic acid, 2-2-methyl-3-nitrophenyl acetic acid, 2-methyl-3-nitro-benzeneacetic acid, 3-nitro-o-tolyl acetic acid, benzeneacetic acid, 2-methyl-3-nitro, 2-methyl-3-nitrophenylaceticacid, 2-methyl-3-nitrobenzeneacetic acid, 2-methyl-3-nitro phenyl acetic acid, pubchem18230
PubChem CID 13530897
SMILES CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitrophenylacetic Acid