missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitroaniline 98.0+%, TCI America™

Catalog No. N045525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N045525G 25 g
N0455250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N045525G Supplier TCI America Supplier No. N045525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 603-83-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007731
InChI Key HFCFJYRLBAANKN-UHFFFAOYSA-N
Synonym 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
PubChem CID 11783
IUPAC Name 2-methyl-3-nitroaniline
SMILES CC1=C(N)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 90°C
Boiling Point 305°C
Quantity 25 g
UN Number 2660
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitroaniline
RTECSNumber XU8200000
TSCA No