missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butenenitrile 75.0+%, TCI America™

Catalog No. p-7132395
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M084025ML 25 mL
M0840500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M084025ML Supplier TCI America Supplier No. M084025ML

Chemical Identifiers

CAS 16529-56-9
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00042634
InChI Key WBAXCOMEMKANRN-UHFFFAOYSA-N
Synonym 3-Cyano-1-butene, 1-Methyl-2-propenyl Cyanide
PubChem CID 27909
IUPAC Name 2-methylbut-3-enenitrile
SMILES CC(C=C)C#N

Specifications

Color Yellow
Boiling Point 124°C
Quantity 25 mL
UN Number 3273
Formula Weight 81.12
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-butenenitrile