missing translation for 'onlineSavingsMsg'
Learn More

2-Méthyl-3-buten-2-ol 97,0+%, TCI America™

Numéro de catalogue. M0178500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M0178500ML 500 mL
M017825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M0178500ML Fournisseur TCI America Code fournisseur M0178500ML

Identifiants chimiques

CAS 115-18-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00004470
InChI Key HNVRRHSXBLFLIG-UHFFFAOYSA-N
Synonym 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
PubChem CID 8257
IUPAC Name 2-méthylbut-3-en-2-ol
SMILES CC(C)(C=C)O

Spécifications

Melting Point -28°C
Color Jaune
Boiling Point 96°C
Quantity 500 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥97.0% (GC)
Physical Form Liquide
Chemical Name or Material 2-Methyl-3-buten-2-ol
EINECSNumber (2)-2393
RTECSNumber EM9472000
TSCA Yes