missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenoxypropionic Acid 98.0+%, TCI America™

Numéro de catalogue. M298425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M298425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M298425G Fournisseur TCI America Code fournisseur M298425G
Il en reste null

Identifiants chimiques

CAS 943-45-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00129939
InChI Key ILPUOPPYSQEBNJ-UHFFFAOYSA-N
Synonym 2-Phenoxyisobutyric Acid
PubChem CID 70343
IUPAC Name 2-methyl-2-phenoxypropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=CC=C1

Spécifications

Melting Point 100°C
Color White-Yellow
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2-phenoxypropionic Acid
RTECSNumber UF6104200
TSCA No