missing translation for 'onlineSavingsMsg'
Learn More

2-Méthyl-2-pentanol 96,0+%, TCI America™

Numéro de catalogue. M11785ML
Modifier l'affichage
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M11785ML 5 mL
M117825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M11785ML Fournisseur TCI America Code fournisseur M11785ML

Identifiants chimiques

CAS 590-36-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004485
InChI Key WFRBDWRZVBPBDO-UHFFFAOYSA-N
Synonym 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l
PubChem CID 11543
IUPAC Name 2-méthylpentane-2-ol
SMILES CCCC(C)(C)O

Spécifications

Color Jaune
Boiling Point 122°C
Quantity 5 mL
UN Number 2560
Formula Weight 102.18
Percent Purity ≥96.0% (GC)
Physical Form Liquide
Chemical Name or Material 2-Methyl-2-pentanol
EINECSNumber (2)-0217
TSCA Yes