missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-nitro-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7132277
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06395G 5 g
M063925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06395G Supplier TCI America Supplier No. M06395G

Chemical Identifiers

CAS 77-49-6
Molecular Formula C4H9NO4
Molecular Weight (g/mol) 135.12
MDL Number MFCD00024812
InChI Key LOTYADDQWWVBDJ-UHFFFAOYSA-N
Synonym 2-methyl-2-nitro-1,3-propanediol, nmpd, 2-nitro-2-methyl-1,3-propanediol, nitroisobutylglycol, 1,3-propanediol, 2-methyl-2-nitro, 2-methyl-2-nitropropanediol, 1,1-dimethylol-1-nitroethane, caswell no. 573n, epa pesticide chemical code 573900, acmc-1basy
PubChem CID 6480
IUPAC Name 2-methyl-2-nitropropane-1,3-diol
SMILES CC(CO)(CO)[N+]([O-])=O

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 135.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2-nitro-1,3-propanediol
RTECSNumber TY8925000
TSCA Yes