missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-butene, ≥99% (GC), MilliporeSigma™ Supelco™
2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.
Supplier: Merck Emd Millipore 6605050ML
Description
Featured Industries: Environmental and PetroleumSpecifications
| 513-35-9 | |
| 0.662 g/mL (at 25°C) | |
| Neat | |
| MFCD00009276 | |
| 35°C to 38°C | |
| Analytical Standard | |
| 70.13 | |
| Analytical Standard |
| 2.4 (vs. air) | |
| -20°C | |
| CH3 CH=C(CH3)2 | |
| -134°C | |
| beta-Isoamylene; Amylene | |
| 50 mL | |
| ≥99% (GC) |