missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187542
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019919 5 mL
111019097 10 mL
111018191 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111019919 Supplier Merck Emd Millipore Supplier No. 660505ML
Only null left

2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.

Featured Industries: Environmental and Petroleum

Specifications

CAS 513-35-9
Vapor Density 2.4 (vs. air)
Density 0.662 g/mL (at 25°C)
Flash Point -20°C
Physical Form Neat
Linear Formula CH3 CH=C(CH3)2
MDL Number MFCD00009276
Melting Point -134°C
Boiling Point 35°C to 38°C
Synonym beta-Isoamylene; Amylene
Type Analytical Standard
Quantity 5 mL
Formula Weight 70.13
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less