missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Methyl-2,4-pentanediol, 98%

Catalog No. AAL03187AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03187AE 100 mL
AAL03187AP 500 mL
AAL031870F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL03187AE Supplier Thermo Scientific Chemicals Supplier No. L03187AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-41-5 | C6H14O2 | 118.176 g/mol

(±)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.

Solubility
Miscible with water, alcohol, ether, fatty acids and lower aliphatic hydrocarbons. Slightly miscible with carbon tetrachloride.

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004547
InChI Key SVTBMSDMJJWYQN-UHFFFAOYSA-N
Synonym hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
PubChem CID 7870
ChEBI CHEBI:62995
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(CC(C)(C)O)O

Specifications

Melting Point -40°C
Density 0.922
Boiling Point 197°C
Flash Point 93°C (199°F)
Odor Weak
Refractive Index 1.427
Quantity 100 mL
Beilstein 1098298
Sensitivity Hygroscopic
Merck Index 14,4710
Solubility Information Miscible with water,alcohol,ether,fatty acids and lower aliphatic hydrocarbons. Slightly miscible with carbon tetrachloride.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material (+/-)-2-Methyl-2,4-pentanediol
Show More Show Less
Hazard Category H227-H315-H319-X
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P260-P264b-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
EINECSNumber 203-489-0
RTECSNumber SA0810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only