missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020407
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020407 1 mL
111018790 5 mL
111018352 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020407 Supplier Merck Emd Millipore Supplier No. 820591MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Methyl-1-propanol can be obtained via hydroformylation of alkenes with fuel gas mixture in particular, carbon monoxide being the main reactant.

Specifications

CAS 78-83-1
Vapor Pressure 8.8 mmHg (0°C)
Vapor Density 2.55 (vs. air)
Density 0.803 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2OH
MDL Number MFCD00004740
Melting Point -108°C
Boiling Point 108°C
Synonym Isobutanol; Isobutyl alcohol
Quantity 1 mL
Formula Weight 74.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less