missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, 99+%, spectrophotometric grade

Catalog No. AC167705000
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167705000 500 mL Glass bottle
AC167700010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167705000 Supplier Thermo Scientific Chemicals Supplier No. 167705000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-83-1 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

CAS Min % 99.0
Absorbance 0.01 max. at 330nm, 0.1 max. at 250nm, 0.5 max. at 230nm, 1 max. at 220nm
CAS Max % 100.0
Melting Point -108.0°C
Density 0.8000g/mL
Boiling Point 108.0°C
Flash Point 28°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3945 to 1.3965
Linear Formula (CH3)2CHCH2OH
Quantity 500 mL
Residue after Evaporation 0.0005% max.
Beilstein 01, 373
Specific Gravity 0.8
Merck Index 15, 5176
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Viscosity 4 Pas (20°C)
Formula Weight 74.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-propanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
EINECSNumber 201-148-0
RTECSNumber NP9625000
TSCA TSCA

RUO – Research Use Only