missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018556
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018556 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018556 Supplier Merck Emd Millipore Supplier No. 6845025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Methyl-1-pentene is a hydrocarbon belonging to the group of volatile organic compounds.

Featured Industries: Environmental and Petroleum

Specifications

CAS 763-29-1
Ignition Point 572°F
Vapor Pressure 325 mmHg (37.7°C)
Density 0.682 g/mL (at 25°C)
Flash Point -27°C - closed cup
Physical Form Neat
Linear Formula CH3 CH2 CH2 C(CH3)=CH2
MDL Number MFCD00009405
Melting Point -136°C
Boiling Point 62°C
Type Analytical Standard
Quantity 25 mL
Formula Weight 84.16
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less