missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentanol 98.0+%, TCI America™

Catalog No. M019225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M019225ML 25 mL
M0192500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M019225ML Supplier TCI America Supplier No. M019225ML
Only null left

Chemical Identifiers

CAS 105-30-6
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004745
InChI Key PFNHSEQQEPMLNI-UHFFFAOYSA-N
Synonym 2-methyl-1-pentanol, 1-pentanol, 2-methyl, 2-methylpentanol-1, 2-methylpentanol, sec-amyl carbinol, 2-methylamyl alcohol, +--2-methylpentanol, 2-mpoh, +--2-methyl-1-pentanol, 2-methyl-2-propylethanol
PubChem CID 7745
IUPAC Name 2-methylpentan-1-ol
SMILES CCCC(C)CO

Specifications

Color Colorless
Boiling Point 148°C
Quantity 25 mL
UN Number 1987
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-pentanol
EINECSNumber (2)-0217
RTECSNumber SA7175000
TSCA Yes