missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-butanethiol 95.0+%, TCI America™

Catalog No. M10415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10415G Supplier TCI America Supplier No. M10415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1878-18-8
Color Yellow
Boiling Point 119°C
Molecular Formula C5H12S
Quantity 5 g
MDL Number MFCD00004883
UN Number 3336
Synonym 2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
InChI Key WGQKBCSACFQGQY-UHFFFAOYNA-N
SMILES CCC(C)CS
IUPAC Name 2-methylbutane-1-thiol
Molecular Weight (g/mol) 104.21
PubChem CID 15877
Formula Weight 104.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-butanethiol
Show More Show Less
EINECSNumber (2)-0464
TSCA Yes