missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-pentadiene 99.0+%, TCI America™

Catalog No. M06161ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06161ML 1 mL
M06165ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06161ML Supplier TCI America Supplier No. M06161ML

Chemical Identifiers

CAS 763-30-4
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008601
InChI Key DRWYRROCDFQZQF-UHFFFAOYSA-N
PubChem CID 12987
IUPAC Name 2-methylpenta-1,4-diene
SMILES CC(=C)CC=C

Specifications

Color Colorless
Boiling Point 57°C
Quantity 1 mL
UN Number 2461
Formula Weight 82.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,4-pentadiene
RTECSNumber RZ2473500
TSCA No