missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-1,3-thiazol-4-yl)methanol, 97%, Thermo Scientific™

Catalog No. CC05709DE
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC05709DE 5g Amber Glass Bottle
CC05709DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC05709DE Supplier Thermo Scientific Chemicals Supplier No. CC05709DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76632-23-0
Molecular Formula C5H7NOS
Molecular Weight (g/mol) 129.177
InChI Key KRCIFTBSQKDYMH-UHFFFAOYSA-N
Synonym 2-methyl-1,3-thiazol-4-yl methanol, 2-methylthiazol-4-yl methanol, 4-hydroxymethyl-2-methylthiazole, 4-thiazolemethanol, 2-methyl, 2-methyl-thiazol-4-yl-methanol, 2-methyl-4-thiazolemethanol, 4-thiazolemethanol,2-methyl, 2-methyl-4-thiazolyl methanol, 4-hydroxymethyl-2-methyl thiazole
PubChem CID 10582809
IUPAC Name (2-methyl-1,3-thiazol-4-yl)methanol
SMILES CC1=NC(=CS1)CO

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 129.18
Percent Purity 97%
Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methanol
Hazard Category GHS Signal Word: Warning