missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M21591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21591G Supplier TCI America Supplier No. M21591G

Specifications

Chemical Name or Material 2-Methoxypyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 163105-90-6
Melting Point 145°C
Color White
Molecular Formula C6H8BNO3
Quantity 1 g
MDL Number MFCD03411572
Synonym 2-methoxypyridine-3-boronic acid, 2-methoxy-3-pyridineboronic acid, 2-methoxypyridin-3-yl boronic acid, 2-methoxy-3-pyridylboronic acid, 2-methoxypyridin-3-yl-3-boronic acid, 2-methoxy-3-pyridinyl boronic acid, 2-methoxy-3-pyridyl boronic acid, 2-methoxy-3-pyridinyl-boronic acid, boronic acid, 2-methoxy-3-pyridinyl, boronic acid, b-2-methoxy-3-pyridinyl
InChI Key NVOLYUXUHWBCRJ-UHFFFAOYSA-N
SMILES COC1=NC=CC=C1B(O)O
IUPAC Name (2-methoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 152.94
PubChem CID 2762709
Formula Weight 152.94
Physical Form Crystalline Powder
Show More Show Less