missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxynaphthalene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 164755000
| Quantity | 500g |
|---|---|
| Packaging | Plastic bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C11H10O | |
| MFCD00004061 | |
| naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether | |
| 2-methoxynaphthalene |
Specifications
| 2-Methoxynaphthalene | |
| 93-04-9 | |
| 100.0 | |
| Authentic | |
| Plastic bottle | |
| C10H7OCH3 | |
| MFCD00004061 | |
| 15, 6070 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,carbon disulfide and,benzene | |
| COC1=CC2=CC=CC=C2C=C1 | |
| 158.2 | |
| 158.2 |
| 98% | |
| 97.5 | |
| 274°C | |
| 98% | |
| C11H10O | |
| 500g | |
| 06, 640 | |
| naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether | |
| LUZDYPLAQQGJEA-UHFFFAOYSA-N | |
| 2-methoxynaphthalene | |
| 7119 | |
| 98% |
Safety and Handling
EINECSNumber : 202-213-6
RTECSNumber : QJ9468750
TSCA : TSCA
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.