missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl isothiocyanate, 98+%

Catalog No. AAL1083906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1083906 5 g
AAL1083903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1083906 Supplier Thermo Scientific Chemicals Supplier No. L1083906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38663-85-3 | C4H7NOS | 117.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38663-85-3
Molecular Formula C4H7NOS
Molecular Weight (g/mol) 117.166
MDL Number MFCD00041126
InChI Key HTSAVXAFEVUJQE-UHFFFAOYSA-N
Synonym 2-methoxyethyl isothiocyanate, ethane, 1-isothiocyanato-2-methoxy, 2-methoxyethylisothiocyanate, 1-isothiocyanato-2-methoxy-ethane, 1-isothiocyanato-2-methoxyethane, 2-methoxyethanisothiocyanate, acmc-1aezw, 2-methoxy ethyl isothiocyanate, #, ethane,1-isothiocyanato-2-methoxy
PubChem CID 123475
IUPAC Name 1-isothiocyanato-2-methoxyethane
SMILES COCCN=C=S

Specifications

Density 1.08
Boiling Point 88°C to 92°C (24 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.512
Quantity 5 g
UN Number UN2922
Beilstein 773990
Sensitivity Moisture sensitive
Formula Weight 117.17
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyethyl isothiocyanate
Show More Show Less
Hazard Category H227-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 254-070-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only