missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. M101725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M101725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M101725G Supplier TCI America Supplier No. M101725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO

Specifications

Color Yellow
Boiling Point 250°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzyl Alcohol