missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxyacetophenone 97.0+%, TCI America™

Catalog No. M144525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M144525G 25 g
M1445250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M144525G Supplier TCI America Supplier No. M144525G

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethan-1-one
SMILES COC1=CC=CC=C1C(C)=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Methoxyacetophenone
EINECSNumber (3)-2676
TSCA Yes