missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxyacetophenone, 98%

Catalog No. AAA1192922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1192922 100 g
AAA1192914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1192922 Supplier Thermo Scientific Chemicals Supplier No. A1192922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 579-74-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(C)=O

Specifications

Density 1.09
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.5393
Quantity 100 g
Beilstein 1100602
Sensitivity Light sensitive
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2'-Methoxyacetophenone
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 209-446-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only